missing translation for 'onlineSavingsMsg'
Learn More

2,4-Hexadienoic acid, 99%, Thermo Scientific Chemicals

Product Code. 10714621
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-44-1
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00002703
InChI Key WSWCOQWTEOXDQX-MQQKCMAXSA-N
Synonym sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid
PubChem CID 643460
ChEBI CHEBI:38358
IUPAC Name (2E,4E)-hexa-2,4-dienoic acid
SMILES C\C=C\C=C\C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 132°C to 135°C
Color Cream-White or White
Flash Point 127°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula CH3CH=CHCH=CHCO2H
Quantity 1 kg
Beilstein 02, 483
Fieser 15,296
Merck Index 15, 8847
Solubility Information Solubility in water: 1.6g/L (20°C). Other solubilities: soluble in ethanol,chloroform,very soluble in diethyl ether
Formula Weight 112.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 4-Hexadienoic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'