missing translation for 'onlineSavingsMsg'
Learn More
2-(4-Hydroxyphenylazo)benzoic acid, 97%, Thermo Scientific Chemicals Product Code.: 11494103

2-(4-Hydroxyphenylazo)benzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 11494103
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11494103

Brand: Thermo Scientific Alfa Aesar A10333.09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Indicator used in determination of serum albumin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-82-8
Molecular Formula C13H10N2O3
Molecular Weight (g/mol) 242.234
MDL Number MFCD00002428
InChI Key FBVSMDPNVYJNON-UHFFFAOYSA-N
Synonym 2-4-hydroxyphenylazo benzoic acid, haba, 2-p-hydroxyphenylazo benzoic acid, benzoic acid, 2-4-hydroxyphenyl azo, o-p-hydroxyphenylazo benzoic acid, 2-4'-hydroxyphenyl-azo benzoic acid, hbaba, 2-4-hydroxyphenyl diazenyl benzoic acid, 2-2-4-hydroxyphenyl diazen-1-yl benzoic acid, benzoic acid, o-p-hydroxyphenyl azoShow MoreShow Less
PubChem CID 5357439
IUPAC Name 2-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)NN=C2C=CC(=O)C=C2

Specifications

Melting Point 205°C to 207°C
Quantity 10 g
Beilstein 1842696
Formula Weight 242.23
Percent Purity ≥98%
Chemical Name or Material 2- (4-Hydroxyphenylazo)benzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10248674 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberA10333.18,A10333.09,A10333.30
Lot NumberB7450A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10333.18,A10333.09
Lot NumberB7450C Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10333.18,A10333.09
Lot Number10211276 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10333.18,A10333.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(4-Hydroxyphenylazo)benzoic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.