Learn More
2-(4-Isobutylphenyl)propanenitrile, 97%, Thermo Scientific™
138.00 € - 565.00 €
Chemical Identifiers
CAS | 58609-73-7 |
---|---|
Molecular Formula | C13H17N |
Molecular Weight (g/mol) | 187.286 |
MDL Number | MFCD02180706 |
InChI Key | PKQKHWNHCKNYSW-UHFFFAOYSA-N |
Synonym | 2-4-isobutylphenyl propanenitrile, 2-4-isobutylphenyl propionitrile, 2-4-2-methylpropyl phenyl propanenitrile, 2-4-isobutylphenyl propiononitrile, 2-p-isobutylphenyl propiononitrile, +--2-4'-isobutylphenyl propionitrile, +/--2-4'-isobutylphenyl propionitrile, ibuprofen nitrile, aronis24266, 2-4-isobutylphenyl-propionitrileShow More |
PubChem CID | 108374 |
ChEBI | CHEBI:27415 |
IUPAC Name | 2-[4-(2-methylpropyl)phenyl]propanenitrile |
SMILES | CC(C)CC1=CC=C(C=C1)C(C)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
10620933
|
Thermo Scientific Maybridge
SEW06499DA |
1 g | N/A | N/A | N/A | |||||
10526435
|
Thermo Scientific Maybridge
SEW06499EA |
10 g |
565.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Chemical Identifiers
58609-73-7 | |
187.286 | |
PKQKHWNHCKNYSW-UHFFFAOYSA-N | |
108374 | |
2-[4-(2-methylpropyl)phenyl]propanenitrile |
C13H17N | |
MFCD02180706 | |
2-4-isobutylphenyl propanenitrile, 2-4-isobutylphenyl propionitrile, 2-4-2-methylpropyl phenyl propanenitrile, 2-4-isobutylphenyl propiononitrile, 2-p-isobutylphenyl propiononitrile, +--2-4'-isobutylphenyl propionitrile, +/--2-4'-isobutylphenyl propionitrile, ibuprofen nitrile, aronis24266, 2-4-isobutylphenyl-propionitrileShow More | |
CHEBI:27415 | |
CC(C)CC1=CC=C(C=C1)C(C)C#N |
Specifications
58609-73-7 | |
97% | |
C13H17N | |
MFCD02180706 | |
PKQKHWNHCKNYSW-UHFFFAOYSA-N | |
2-[4-(2-methylpropyl)phenyl]propanenitrile | |
108374 | |
187.28 | |
2-(4-Isobutylphenyl)propanenitrile |
97.0 | |
Amber Glass Bottle | |
(CH3)2CHCH2C6H4CH(CN)CH3 | |
2-4-isobutylphenyl propanenitrile, 2-4-isobutylphenyl propionitrile, 2-4-2-methylpropyl phenyl propanenitrile, 2-4-isobutylphenyl propiononitrile, 2-p-isobutylphenyl propiononitrile, +--2-4'-isobutylphenyl propionitrile, +/--2-4'-isobutylphenyl propionitrile, ibuprofen nitrile, aronis24266, 2-4-isobutylphenyl-propionitrile | |
CC(C)CC1=CC=C(C=C1)C(C)C#N | |
187.286 | |
CHEBI:27415 | |
97% |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.