missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylbenzylamino)pyridine-5-boronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 15487685
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073354-32-1
Molecular Formula C19H25BN2O2
Molecular Weight (g/mol) 324.231
MDL Number MFCD06798272
InChI Key AFMJUGQUPACTDK-UHFFFAOYSA-N
Synonym n-4-methylbenzyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 6-4-methylbenzylamino pyridine-3-boronic acid pinacol ester, 2-4-methylbenzylamino pyridine-5-boronic acid pinacol ester, n-4-methylbenzyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinamine, n-4-methylphenyl methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, n-4-methylbenzyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2-pyridinamine, n-4-methylphenyl methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 2-pyridinamine, n-4-methylphenyl methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, n-4-methylphenyl methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinamine
PubChem CID 44755166
IUPAC Name N-[(4-methylphenyl)methyl]-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)NCC3=CC=C(C=C3)C

Specifications

Melting Point 148°C to 153°C
Quantity 250 mg
Formula Weight 324.23
Percent Purity 95%
Chemical Name or Material 2-(4-Methylbenzylamino)pyridine-5-boronic acid pinacol ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.