missing translation for 'onlineSavingsMsg'
Learn More

2,4-Pentanedione, 99%, Thermo Scientific Chemicals

27.74 € - 148.74 €

Chemical Identifiers

CAS 123-54-6
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00008787
InChI Key YRKCREAYFQTBPV-UHFFFAOYSA-N
Synonym 2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
PubChem CID 31261
ChEBI CHEBI:14750
IUPAC Name pentane-2,4-dione
SMILES CC(=O)CC(C)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11454080
View Documents
Thermo Scientific Alfa Aesar
A14117.AE
100 mL
27.74 €
100mL
Please to purchase this item. Need a web account? Register with us today!
11488076
View Documents
Thermo Scientific Alfa Aesar
A14117.AP
500 mL
41.91 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11498076
View Documents
Thermo Scientific Alfa Aesar
A14117.0F
2500 mL
148.74 €
2500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2,4-Pentanedione is used as a precursor to acetylacetonate, a common bidentate and a raw material for the synthesis of heterocyclic compounds. It is used in pharmaceuticals, organic intermediates for solvents, analytical reagents, tungsten and molybdenum in aluminum extraction agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Pentanedione is used as a precursor to acetylacetonate, a common bidentate and a raw material for the synthesis of heterocyclic compounds. It is used in pharmaceuticals, organic intermediates for solvents, analytical reagents, tungsten and molybdenum in aluminum extraction agent.

Solubility
It is readily miscible with water.

Notes
Incompatible with isocyanates, aldehydes, cyanides, peroxides, and anhydrides.
Specifications

Chemical Identifiers

123-54-6
100.12
YRKCREAYFQTBPV-UHFFFAOYSA-N
31261
pentane-2,4-dione
C5H8O2
MFCD00008787
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
CHEBI:14750
CC(=O)CC(C)=O

Specifications

123-54-6
0.973
34°C (93°F)
C5H8O2
MFCD00008787
741937
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone
YRKCREAYFQTBPV-UHFFFAOYSA-N
pentane-2,4-dione
31261
100.12
Liquid
-23°C
140°C to 141°C
99%
1.452
UN2310
14,81
It is readily miscible with water.
CC(=O)CC(C)=O
100.12
CHEBI:14750
99%
2,4-Pentanedione
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H311-H331-H226-H302
Toxic in contact with skin.
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P311-P312-P330-P363-P370+P378q-P501c

H226-H302-H311+H331

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANE-2,4-DIONE

missing translation for 'einecsNumber' : 204-634-0

missing translation for 'rtecsNumber' : SA1925000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • 2,4-Pentanedione

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : 2,4-Pentanedione

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only