Learn More
2-(4-Piperidinyloxy)benzonitrile, 98%, Thermo Scientific™
72.40 € - 262.00 €
Chemical Identifiers
CAS | 900572-37-4 |
---|---|
Molecular Formula | C12H14N2O |
Molecular Weight (g/mol) | 202.26 |
MDL Number | MFCD08061090 |
InChI Key | OIFRRCRLKBGBKR-UHFFFAOYSA-N |
Synonym | 2-4-piperidinyloxy benzonitrile, 2-piperidin-4-yloxy benzonitrile, acmc-20aoor, 2-4-piperidyloxy benzenecarbonitrile, 2-piperidin-4-yl oxy benzonitrile, benzonitrile, 2-4-piperidinyloxy, 2-piperidin-4-yl-oxy-benzonitrile |
PubChem CID | 16732591 |
IUPAC Name | 2-piperidin-4-yloxybenzonitrile |
SMILES | N#CC1=CC=CC=C1OC1CCNCC1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
900572-37-4 | |
202.26 | |
OIFRRCRLKBGBKR-UHFFFAOYSA-N | |
16732591 | |
N#CC1=CC=CC=C1OC1CCNCC1 |
C12H14N2O | |
MFCD08061090 | |
2-4-piperidinyloxy benzonitrile, 2-piperidin-4-yloxy benzonitrile, acmc-20aoor, 2-4-piperidyloxy benzenecarbonitrile, 2-piperidin-4-yl oxy benzonitrile, benzonitrile, 2-4-piperidinyloxy, 2-piperidin-4-yl-oxy-benzonitrile | |
2-piperidin-4-yloxybenzonitrile |
Specifications
900572-37-4 | |
MFCD08061090 | |
2-4-piperidinyloxy benzonitrile, 2-piperidin-4-yloxy benzonitrile, acmc-20aoor, 2-4-piperidyloxy benzenecarbonitrile, 2-piperidin-4-yl oxy benzonitrile, benzonitrile, 2-4-piperidinyloxy, 2-piperidin-4-yl-oxy-benzonitrile | |
N#CC1=CC=CC=C1OC1CCNCC1 | |
202.26 | |
202.26 | |
2-(4-Piperidinyloxy)benzonitrile |
C12H14N2O | |
UN3439 | |
OIFRRCRLKBGBKR-UHFFFAOYSA-N | |
2-piperidin-4-yloxybenzonitrile | |
16732591 | |
98% |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures