missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Pyridyl)-1H-anthra[1,2-d]imidazole-6,11-dione, 97%, Thermo Scientific™

Product Code. 15431766
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1347815-30-8
Molecular Formula C20H11N3O2
Molecular Weight (g/mol) 325.327
MDL Number MFCD20265373
InChI Key YZNTXTFZKAVVRN-UHFFFAOYSA-N
Synonym 2-pyridin-4-yl-1h-anthra 1,2-d imidazole-6,11-dione
PubChem CID 73996030
IUPAC Name 2-pyridin-4-yl-3H-naphtho[3,2-e]benzimidazole-6,11-dione
SMILES C1=CC=C2C(=C1)C(=O)C3=C(C2=O)C4=C(C=C3)NC(=N4)C5=CC=NC=C5

Specifications

Quantity 1 g
Formula Weight 325.33
Percent Purity 97%
Chemical Name or Material 2-(4-Pyridyl)-1H-anthra[1,2-d]imidazole-6,11-dione
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'