missing translation for 'onlineSavingsMsg'
Learn More
2,5-Bis(4-biphenylyl)-1,3,4-oxadiazole, 99%, Thermo Scientific Chemicals Product Code.: 11342087

2,5-Bis(4-biphenylyl)-1,3,4-oxadiazole, 99%, Thermo Scientific Chemicals

Product Code. 11342087
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 11342087

Brand: Thermo Scientific Alfa Aesar L07874.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2043-06-3
MDL Number MFCD00042666

Specifications

Melting Point 235°C to 238°C
Molecular Formula C26H18N2O
Quantity 1 g
Beilstein 318611
Synonym 2,5-bis 4-biphenylyl-1,3,4-oxadiazole, 2,5-bis 4-phenylphenyl-1,3,4-oxadiazole, 2,5-bis 1,1'-biphenyl-4-yl-1,3,4-oxadiazole, acmc-20aobr, 2,5-bis 4-biphenylyl oxadiazole, 2,5-bis biphenyl-4-yl-1,3,4-oxadiazole, 2,5-di-4-biphenylyl-1,3,4-oxadiazole, 2,5_di-4-biphenylyl-1,3,4-oxadiazole, 2,5-bis-4-biphenyl-yl-1,3,4-oxadiazole
InChI Key IVVYVRLKDGGUEL-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=NN=C(O3)C4=CC=C(C=C4)C5=CC=CC=C5
IUPAC Name 2,5-bis(4-phenylphenyl)-1,3,4-oxadiazole
Molecular Weight (g/mol) 374.44
PubChem CID 96840
Formula Weight 374.45
Percent Purity 99%
Chemical Name or Material 2,5-Bis(4-biphenylyl)-1,3,4-oxadiazole
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Bis(4-biphenylyl)-1,3,4-oxadiazole, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.