missing translation for 'onlineSavingsMsg'
Learn More
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%, Thermo Scientific Chemicals Product Code.: 11472747

2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%, Thermo Scientific Chemicals

Product Code. 11472747
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11472747

Brand: Thermo Scientific Alfa Aesar A14928.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene is used as an optical brightener which converts UV light into visible light. It is also used as a fluorescent brightener for thermoplastic resins of polyvinyl chloride, acrylic resin, polyester fiber, paint, coating and printing ink. It is also useful in detecting the presence of thin-film electrical contact lubricants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene is used as an optical brightener which converts UV light into visible light. It is also used as a fluorescent brightener for thermoplastic resins of polyvinyl chloride, acrylic resin, polyester fiber, paint, coating and printing ink. It is also useful in detecting the presence of thin-film electrical contact lubricants.

Solubility
Soluble in acetone and toluene. Insoluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7128-64-5
Molecular Formula C26H26N2O2S
Molecular Weight (g/mol) 430.566
MDL Number MFCD00005774
InChI Key AIXZBGVLNVRQSS-UHFFFAOYSA-N
Synonym bbot, 2,5-bis 5-tert-butyl-2-benzoxazolyl thiophene, uvitex ob, 2,5-bis 5-tert-butyl benzo d oxazol-2-yl thiophene, bbot 150, 2,5-bis 5-tert-butylbenzoxazol-2-yl thiophene, benzoxazole, 2,2'-2,5-thiophenediyl bis 5-1,1-dimethylethyl, unii-w7ur9129gp, fluorescent brightener 184, 2,2'-2,5-thiophenediyl bis 5-tert-butylbenzoxazoleShow MoreShow Less
PubChem CID 292429
IUPAC Name 5-tert-butyl-2-[5-(5-tert-butyl-1,3-benzoxazol-2-yl)thiophen-2-yl]-1,3-benzoxazole
SMILES CC(C)(C)C1=CC2=C(C=C1)OC(=N2)C3=CC=C(S3)C4=NC5=C(O4)C=CC(=C5)C(C)(C)C

Specifications

Melting Point 200°C to 202°C
Quantity 5 g
Beilstein 631803
Solubility Information Soluble in acetone and toluene. Insoluble in water.
Formula Weight 430.57
Percent Purity 99%
Chemical Name or Material 2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252053 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog NumberA14928.06,A14928.14,A14928.22
Lot Number10248966 Certificate TypeCertificate of Analysis Date13/12/2023 Catalog NumberA14928.06,A14928.14,A14928.22
Lot Number10237844 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14928.06,A14928.14,A14928.22
Lot Number10230380 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14928.06,A14928.14,A14928.22
Lot Number10196840 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14928.06,A14928.14,A14928.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Bis(5-tert-butyl-2-benzoxazolyl)thiophene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.