missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals 2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol, 98%

Product Code. 10742475 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10742475

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14337-53-2 | C15H17BrN4O | 349.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14337-53-2
Molecular Formula C15H17BrN4O
Molecular Weight (g/mol) 349.23
MDL Number MFCD00006255
InChI Key LYVSIKOGJUDRBK-QGOAFFKASA-N
Synonym 2-5-bromo-2-pyridylazo-5-diethylamino phenol, 2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol, bromo-padap, 2-5-bromo-2-pyridylazo-5-diethylaminophenol, phenol, 2-5-bromo-2-pyridinyl azo-5-diethylamino, 2-4-diethylamino-2-hydroxyphenylazo-5-bromopyridine, hnvcxdavehoibp-vhebqxmusa-n, 2-5-bromo-2-pyridylazo-5-diethylamino-phenol, 5-diethylamino-2-5-bromo 2-pyridyl diazenyl phenol, e-2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol
PubChem CID 5911417
IUPAC Name (6E)-6-[(5-bromopyridin-2-yl)hydrazinylidene]-3-(diethylamino)cyclohexa-2,4-dien-1-one
SMILES CCN(CC)C1=CC(=O)\C(C=C1)=N\NC1=CC=C(Br)C=N1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 155.0°C to 160.0°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 349.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.