missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Chlorobenzo[b]thiophen-3-yl)acetic acid, 97%, Thermo Scientific™

53.10 € - 145.00 €

Chemical Identifiers

CAS 17266-30-7
Molecular Formula C10H7ClO2S
Molecular Weight (g/mol) 226.674
MDL Number MFCD00052308
InChI Key QQKKTOPRRGBBCT-UHFFFAOYSA-N
Synonym 5-chlorobenzo b thiophene-3-acetic acid, 2-5-chlorobenzo b thiophen-3-yl acetic acid, 2-5-chloro-1-benzothiophen-3-yl acetic acid, benzo b thiophene-3-acetic acid, 5-chloro, 5-chloro-1-benzothien-3-yl acetic acid, 5-chloro-1-benzothiophen-3-yl acetic acid, maybridge4_002620, acmc-20am45
PubChem CID 205058
IUPAC Name 2-(5-chloro-1-benzothiophen-3-yl)acetic acid
SMILES C1=CC2=C(C=C1Cl)C(=CS2)CC(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10551112
View Documents
Thermo Scientific Maybridge
MWP00086CB
250 mg
53.10 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10446282
View Documents
Thermo Scientific Maybridge
MWP00086DA
1 g
145.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

17266-30-7
226.674
QQKKTOPRRGBBCT-UHFFFAOYSA-N
205058
C1=CC2=C(C=C1Cl)C(=CS2)CC(=O)O
C10H7ClO2S
MFCD00052308
5-chlorobenzo b thiophene-3-acetic acid, 2-5-chlorobenzo b thiophen-3-yl acetic acid, 2-5-chloro-1-benzothiophen-3-yl acetic acid, benzo b thiophene-3-acetic acid, 5-chloro, 5-chloro-1-benzothien-3-yl acetic acid, 5-chloro-1-benzothiophen-3-yl acetic acid, maybridge4_002620, acmc-20am45
2-(5-chloro-1-benzothiophen-3-yl)acetic acid

Specifications

17266-30-7
97%
MFCD00052308
QQKKTOPRRGBBCT-UHFFFAOYSA-N
2-(5-chloro-1-benzothiophen-3-yl)acetic acid
205058
97%
97.0
C10H7ClO2S
5-chlorobenzo b thiophene-3-acetic acid, 2-5-chlorobenzo b thiophen-3-yl acetic acid, 2-5-chloro-1-benzothiophen-3-yl acetic acid, benzo b thiophene-3-acetic acid, 5-chloro, 5-chloro-1-benzothien-3-yl acetic acid, 5-chloro-1-benzothiophen-3-yl acetic acid, maybridge4_002620, acmc-20am45
C1=CC2=C(C=C1Cl)C(=CS2)CC(=O)O
226.674
226.68
2-(5-Chlorobenzo[b]thiophen-3-yl)acetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications