missing translation for 'onlineSavingsMsg'
Learn More
2,5-Diaminohydroquinone dihydrochloride, 97%, Thermo Scientific Chemicals Product Code.: 11425087

2,5-Diaminohydroquinone dihydrochloride, 97%, Thermo Scientific Chemicals

Product Code. 11425087
1 g
Click to view available options
Quantity:
0.25 g
1 g
This item is not returnable. View return policy

Product Code. 11425087

Brand: Thermo Scientific Alfa Aesar B21969.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,5-Diaminohydroquinone dihydrochloride is used in the preparation of polymers, synthetically cross-linked polyimide and silica hybrid films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Diaminohydroquinone dihydrochloride is used in the preparation of polymers, synthetically cross-linked polyimide and silica hybrid films.

Notes
Air sensitive. It may discolor to black-violet on storage. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24171-03-7
Molecular Formula C6H10Cl2N2O2
Molecular Weight (g/mol) 213.058
MDL Number MFCD00239416
InChI Key NILKAWPWTYPHAH-UHFFFAOYSA-N
Synonym 2,5-diaminobenzene-1,4-diol dihydrochloride, 2,5-diamino-1,4-dihydroxybenzene dihydrochloride, 2,5-diaminohydroquinone dihydrochloride, acmc-20aobi, 1,4-benzenediol, 2,5-diamino-, dihydrochloride, 2,5-diaminobenzene-1,4-diol, chloride, chloride, 1,4-benzenediol,2,5-diamino-,hydrochloride 1:2, 2,5-diaminobenzene-1,4-diol-hydrogen chloride 1/2, 2,5-diaminohydroquinone dihydrochloride, technical at
PubChem CID 16212088
IUPAC Name 2,5-diaminobenzene-1,4-diol;dihydrochloride
SMILES C1=C(C(=CC(=C1O)N)O)N.Cl.Cl

Specifications

Melting Point >300°C
Odor Odorless
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 213.06
Percent Purity 97%
Chemical Name or Material 2,5-Diaminohydroquinone dihydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10223285 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB21969.01,B21969.03
Lot Number10232794 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB21969.01,B21969.03
Lot Number10222160 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21969.01,B21969.03
Lot Number10209818 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21969.01,B21969.03
Lot Number10213210 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21969.01,B21969.03
compliance-icons
Product Identifier
  • 2,5-Diamino-1,4-dihydroxybenzenedihydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,5-Diaminohydroquinone dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Diaminohydroquinone dihydrochloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.