missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dibromoaniline, 97%, Thermo Scientific Chemicals Product Code.: 10296100

2,5-Dibromoaniline, 97%, Thermo Scientific Chemicals

Product Code. 10296100
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10296100

Brand: Thermo Scientific Acros 155030250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3638-73-1
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.92
MDL Number MFCD00007636
InChI Key WRTAZRGRFBCKBU-UHFFFAOYSA-N
Synonym 2,5-dibromobenzenamine, benzenamine, 2,5-dibromo, 2,5-dibromophenylamine, rarechem fh 1w 0043, pubchem3452, 2,5-dibromo-phenylamine, aniline, 2,5-dibromo, 2,5-bis bromanyl aniline, acmc-209im6, 2,5-dibromoaniline
PubChem CID 77198
IUPAC Name 2,5-dibromoaniline
SMILES C1=CC(=C(C=C1Br)N)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 51.0°C to 56.0°C
Color Beige to Gray
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula Br2C6H3NH2
Quantity 25 g
Beilstein 12, 659
Formula Weight 250.92
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Dibromoaniline, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0432474 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog Number155030050,155030250
Lot NumberA0371827 Certificate TypeCertificate of Analysis Date23/10/2023 Catalog Number155030050,155030250
compliance-icons compliance-icons
Product Identifier
  • 2,5-Dibromoaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dibromoaniline, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.