missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dibromobenzotrifluoride, 98%, Thermo Scientific™ Product Code.: 11352126

2,5-Dibromobenzotrifluoride, 98%, Thermo Scientific™

Product Code. 11352126
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11352126

Brand: Thermo Scientific Alfa Aesar L03378.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7657-09-2
MDL Number MFCD00013554
ChEBI CHEBI:27656

Specifications

Melting Point 47°C to 51°C
Boiling Point 210°C
Flash Point >100°C (212°F)
Molecular Formula C20H16N2O4
Quantity 5 g
Beilstein 2261093
Synonym camptothecin, camptothecine, s-+-camptothecin, campathecin, +-camptothecine, d-camptothecin, 20 s-camptothecine, +-camptothecin, 21,22-secocamptothecin-21-oic acid lactone, s-camptothecin
InChI Key VSJKWCGYPAHWDS-FQEVSTJZSA-N
SMILES CCC1(C2=C(COC1=O)C(=O)N3CC4=CC5=CC=CC=C5N=C4C3=C2)O
Molecular Weight (g/mol) 348.36
PubChem CID 24360
Formula Weight 303.91
Percent Purity 98%
Chemical Name or Material 2,5-Dibromobenzotrifluoride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberUSLF000333 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number90012881 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10137480 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberB17W029 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10145605 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dibromobenzotrifluoride, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.