missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dibromothiophene, 95%, Thermo Scientific Chemicals Product Code.: 11434080

2,5-Dibromothiophene, 95%, Thermo Scientific Chemicals

Product Code. 11434080
25 g
Click to view available options
Quantity:
25 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11434080

Brand: Thermo Scientific Alfa Aesar A13406.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dibromothiophene, is mainly used as pharmaceutical intermediate and synthesis intermediate.

Solubility
It is soluble in chloroform and methanol. It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.Light Sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3141-27-3
Molecular Formula C4H2Br2S
Molecular Weight (g/mol) 241.93
MDL Number MFCD00005420
InChI Key KBVDUUXRXJTAJC-UHFFFAOYSA-N
Synonym thiophene, 2,5-dibromo, 2,5-dibromo-thiophene, unii-f0033k8cxm, pubchem5512, thiophene,5-dibromo, acmc-1ckmh, 2,5-bis bromanyl thiophene, ksc224g0j, 2,5-dibromothiophene, bidd:gt0086Show MoreShow Less
PubChem CID 18453
IUPAC Name 2,5-dibromothiophene
SMILES BrC1=CC=C(Br)S1

Specifications

Melting Point -6°C
Density 2.15
Boiling Point 210°C to 212°C
Flash Point 99°C (210°F)
Odor Pungent
Refractive Index 1.629
Quantity 25 g
UN Number UN2810
Beilstein 109899
Solubility Information It is soluble in chloroform and methanol. It is insoluble in water.
Formula Weight 241.94
Percent Purity 96%
Chemical Name or Material 2,5-Dibromothiophene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW12J028 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog NumberA13406.30,A13406.14,A13406.18
Lot Number10233620 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13406.30,A13406.14,A13406.18
Lot NumberH29Q13 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13406.30,A13406.14,A13406.18
Lot Number10154406 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13406.30,A13406.14,A13406.18
Lot NumberH5376A Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13406.30,A13406.14,A13406.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dibromothiophene, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.