missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',5'-Dichloroacetophenone, 96%, Thermo Scientific™
40.00 € - 294.00 €
Chemical Identifiers
CAS | 2476-37-1 |
---|---|
Molecular Formula | C8H6Cl2O |
Molecular Weight (g/mol) | 189.04 |
MDL Number | MFCD00000607 |
InChI Key | CYNFEPKQDJHIMV-UHFFFAOYSA-N |
Synonym | 2',5'-dichloroacetophenone, 1-2,5-dichlorophenyl ethanone, 2,5-dichloroacetophenone, ethanone, 1-2,5-dichlorophenyl, acetophenone, 2',5'-dichloro, 1-2,5-dichlorophenyl ethan-1-one, 1-acetyl-2,5-dichlorobenzene, 2,5-dichloracetophenon, pubchem10586, acmc-1cmfm |
PubChem CID | 75587 |
IUPAC Name | 1-(2,5-dichlorophenyl)ethanone |
SMILES | CC(=O)C1=C(C=CC(=C1)Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15177412
|
Thermo Scientific Acros
179420100 |
10 g | Glass bottle | N/A | |||||
10307650
|
Thermo Scientific Acros
179420500 |
50 g | Glass bottle | N/A | |||||
15187412
|
Thermo Scientific Acros
179421000 |
100 g | Glass bottle | N/A | |||||
Description
Chemical Identifiers
2476-37-1 | |
189.04 | |
CYNFEPKQDJHIMV-UHFFFAOYSA-N | |
75587 | |
CC(=O)C1=C(C=CC(=C1)Cl)Cl |
C8H6Cl2O | |
MFCD00000607 | |
2',5'-dichloroacetophenone, 1-2,5-dichlorophenyl ethanone, 2,5-dichloroacetophenone, ethanone, 1-2,5-dichlorophenyl, acetophenone, 2',5'-dichloro, 1-2,5-dichlorophenyl ethan-1-one, 1-acetyl-2,5-dichlorobenzene, 2,5-dichloracetophenon, pubchem10586, acmc-1cmfm | |
1-(2,5-dichlorophenyl)ethanone |
Specifications
2476-37-1 | |
100.0 | |
Brown to Yellow | |
118.0°C (12.0 mmHg) | |
Authentic | |
C8H6Cl2O | |
1.5604 to 1.5624 | |
1.312 | |
CYNFEPKQDJHIMV-UHFFFAOYSA-N | |
1-(2,5-dichlorophenyl)ethanone | |
75587 | |
96% | |
2', 5'-Dichloroacetophenone, 96% |
95.0 | |
11.0°C to 13.0°C | |
1.3120g/mL | |
>110°C | |
95% min. (GC) | |
Cl2C6H3COCH3 | |
MFCD00000607 | |
2',5'-dichloroacetophenone, 1-2,5-dichlorophenyl ethanone, 2,5-dichloroacetophenone, ethanone, 1-2,5-dichlorophenyl, acetophenone, 2',5'-dichloro, 1-2,5-dichlorophenyl ethan-1-one, 1-acetyl-2,5-dichlorobenzene, 2,5-dichloracetophenon, pubchem10586, acmc-1cmfm | |
CC(=O)C1=C(C=CC(=C1)Cl)Cl | |
189.04 | |
189.04 | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 219-605-8
missing translation for 'tsca' : TSCA
Product Identifier
- 2',5'-Dichloroacetophenone
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title