missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dichloroaniline, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
500 g
2500 g
Description
2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.
Solubility
Soluble in dilute hydrochloric acid.
Notes
Air & Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.
Solubility
Soluble in dilute hydrochloric acid.
Notes
Air & Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Melting Point | 48°C to 52°C |
Boiling Point | 250°C to 251°C |
Flash Point | 139°C (282°F) |
Quantity | 250 g |
UN Number | UN3442 |
Beilstein | 1447438 |
Formula Weight | 162.02 |
Percent Purity | 99% |
Chemical Name or Material | 2,5-Dichloroaniline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'