missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 10676942
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-79-3
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.01
MDL Number MFCD00002416
InChI Key QVTQYSFCFOGITD-UHFFFAOYSA-N
Synonym benzoic acid, 2,5-dichloro, unii-kp7cmr0gsd, 2,5-dichlorobenzoicacid, kp7cmr0gsd, 2,5-dichloro-benzoic acid, ccris 8609, pubchem3685, 2,5dichlorobenzoic acid, wln: qvr bg eg, dsstox_cid_4978
PubChem CID 5784
IUPAC Name 2,5-dichlorobenzoic acid
SMILES OC(=O)C1=CC(Cl)=CC=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 152.0°C to 157.0°C
Color White to Beige
Boiling Point 301.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula Cl2C6H3CO2H
Quantity 250 g
Beilstein 09, 342
Solubility Information Solubility in water: 0.8g/L (14°C). Other solubilities: soluble in most polar solvents,insoluble in non-polar solvents
Formula Weight 191.01
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2, 5-Dichlorobenzoic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dichlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'