missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzyl alcohol, 99%, Thermo Scientific Chemicals

40.60 € - 130.65 €

Chemical Identifiers

CAS 34145-05-6
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00004607
InChI Key LCEIGNVIDJNUGF-UHFFFAOYSA-N
PubChem CID 118604
IUPAC Name (2,5-dichlorophenyl)methanol
SMILES C1=CC(=C(C=C1Cl)CO)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11312037
View Documents
Thermo Scientific Alfa Aesar
L07734.06
5 g
40.60 €
5g
Please to purchase this item. Need a web account? Register with us today!
11322037
View Documents
Thermo Scientific Alfa Aesar
L07734.14
25 g
130.65 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

34145-05-6
177.024
LCEIGNVIDJNUGF-UHFFFAOYSA-N
(2,5-dichlorophenyl)methanol
C7H6Cl2O
MFCD00004607
118604
C1=CC(=C(C=C1Cl)CO)Cl

Specifications

34145-05-6
C7H6Cl2O
3240770
LCEIGNVIDJNUGF-UHFFFAOYSA-N
(2,5-dichlorophenyl)methanol
118604
99%
76°C to 80°C
MFCD00004607
2,5-dichlorophenyl methanol, 2,5-dichlorobenzyl alcohol, benzenemethanol, 2,5-dichloro, 2,5-dichlorobenzylic alcohol, 2,5-dichloro-phenyl-methanol, 2,5-dichlorobenzylalcohol, acmc-1ad0a, 2,5-dichloro benzylalcohol, 2,5-dichloro-benzyl alcohol, benzenemethanol,2,5-dichloro
C1=CC(=C(C=C1Cl)CO)Cl
177.024
177.03
2,5-Dichlorobenzyl alcohol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

missing translation for 'einecsNumber' : 251-850-6

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only