missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorothiophene-3-carbonyl chloride, 95%, Thermo Scientific™

Product Code. 10734003
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 57248-14-3
Molecular Formula C5HCl3OS
Molecular Weight (g/mol) 215.472
MDL Number MFCD00041441
InChI Key YGDFSMANOSKQRU-UHFFFAOYSA-N
Synonym 2,5-dichlorothiophene-3-carbonylchloride, 2,5-dichloro-3-thiophenecarbonyl chloride, 3-thiophenecarbonylchloride, 2,5-dichloro, pubchem15832, 2,5-dichloro-3-thenoyl chloride, 2,5-dichloro-3-thiophenecarbonylchloride, 2,5-dichloro-thiophene-3-carbonyl chloride, 2,5-bis chloranyl thiophene-3-carbonyl chloride
PubChem CID 2777084
IUPAC Name 2,5-dichlorothiophene-3-carbonyl chloride
SMILES C1=C(SC(=C1C(=O)Cl)Cl)Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 215.49
Percent Purity 95%
Chemical Name or Material 2, 5-Dichlorothiophene-3-carbonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'