missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorothiophene-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11362526
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56946-83-9
Molecular Formula C4HCl3O2S2
Molecular Weight (g/mol) 251.52
MDL Number MFCD00051665
InChI Key JJKSHSHZJOWSEC-UHFFFAOYSA-N
Synonym 2,5-dichlorothiophene-3-sulphonyl chloride, 2,5-dichloro-3-thiophenesulfonyl chloride, 3-thiophenesulfonyl chloride, 2,5-dichloro, 2,5-dichlorothiophene-3-sulfonic acid chloride, pubchem7412, 2,5-dichlorothiophene-3-sulfonylchloride, acmc-1avwk, 2,5-dichloro 3-thienyl chlorosulfone, 2,5-dichloro-3-thienylsulfonyl chloride, 2,5-dichlorothien-3-yl-sulfonyl chloride
PubChem CID 2736094
IUPAC Name 2,5-dichlorothiophene-3-sulfonyl chloride
SMILES C1=C(SC(=C1S(=O)(=O)Cl)Cl)Cl

Specifications

Density 1.697
Boiling Point 256°C to 257°C
Flash Point >110°C (230°F)
Refractive Index 1.597
Quantity 1 g
UN Number UN3265
Beilstein 1285315
Sensitivity Moisture sensitive
Formula Weight 251.54
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorothiophene-3-sulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.