missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorotoluene, 98%, Thermo Scientific Chemicals

Product Code. 11489963
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19398-61-9
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.025
MDL Number MFCD00000609
InChI Key KFAKZJUYBOYVKA-UHFFFAOYSA-N
Synonym 2,5-dichlorotoluene, benzene, 1,4-dichloro-2-methyl, toluene, 2,5-dichloro, 1,4-dichloro-2-methyl-benzene, toluene,5-dichloro, pubchem3700, acmc-209exk, ksc490m7d, benzene,4-dichloro-2-methyl
PubChem CID 29572
IUPAC Name 1,4-dichloro-2-methylbenzene
SMILES CC1=C(C=CC(=C1)Cl)Cl

Specifications

Melting Point 4°C to 5°C
Density 1.256
Boiling Point 198°C to 200°C
Flash Point 79°C (174°F)
Refractive Index 1.547
Quantity 100 g
UN Number UN3082
Beilstein 1859112
Formula Weight 161.03
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorotoluene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'