missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dihydrofuran, 98%, Thermo Scientific Chemicals Product Code.: 10341150

2,5-Dihydrofuran, 98%, Thermo Scientific Chemicals

Product Code. 10341150
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10341150

Brand: Thermo Scientific Acros 244780250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1708-29-8
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
InChI Key ARGCQEVBJHPOGB-UHFFFAOYSA-N
Synonym furan, 2,5-dihydro, 3-oxolene, 1-oxa-3-cyclopentene, 2,5-dihydro-furan, furan,5-dihydro, oxa-3-cyclopentene, 2,5 dihydrofurane, 2,5-dihydro-fura, pubchem3987, acmc-1buc6
PubChem CID 15570
IUPAC Name 2,5-dihydrofuran
SMILES C1C=CCO1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 66.0°C to 67.0°C
Flash Point −016°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4310 to 1.4330 (20°C, 589nm)
Quantity 25 g
Specific Gravity 0.92
Solubility Information Solubility in water: immiscible.
Formula Weight 70.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,5-Dihydrofuran
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0424836 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number244780250,244781000
Lot NumberA0397944 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number244780250,244781000
Lot NumberA0407162 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number244780250,244781000
Lot NumberA0446835 Certificate TypeCertificate of Analysis Date27/10/2022 Catalog Number244780250,244781000
Lot NumberA0420162 Certificate TypeCertificate of Analysis Date24/06/2020 Catalog Number244780250,244781000
compliance-icons
Product Identifier
  • 2,5-Dihydrofuran
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dihydrofuran, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.