Learn More
2,5-Dihydroxy-1,4-benzoquinone, 98%, Thermo Scientific Chemicals
30.55 € - 119.60 €
Chemical Identifiers
CAS | 615-94-1 |
---|---|
Molecular Formula | C6H4O4 |
Molecular Weight (g/mol) | 140.09 |
MDL Number | MFCD00001598 |
InChI Key | QFSYADJLNBHAKO-UHFFFAOYSA-N |
Synonym | 2,5-dihydroxy-1,4-benzoquinone, 2,5-dihydroxy-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dihydroxy, p-benzoquinone, 2,5-dihydroxy, 2,5-dihydroxybenzoquinone, 4lbp, 2,5-dihydroxy-p-quinone, 2,5-dihydroxy benzoquinone, 2,5-dhbqop |
PubChem CID | 69213 |
IUPAC Name | 2,5-dihydroxycyclohexa-2,5-diene-1,4-dione |
SMILES | OC1=CC(=O)C(O)=CC1=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11464070
|
Thermo Scientific Alfa Aesar
A11442.06 |
5 g |
30.55 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474070
|
Thermo Scientific Alfa Aesar
A11442.14 |
25 g |
71.66 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430303
|
Thermo Scientific Alfa Aesar
A11442.18 |
50 g |
119.60 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,5-dihydroxy-1,4-benzoquinone dianion, and squarate as bridging ligands in the synthesis and magnetic properties of binuclear iron(III) complexes with oxalate. A Dinuclear 2,5-Dihydroxy-1,4-benzoquinone acts as a bridging ligand Ruthenium(II) Complex with the Dianion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,5-dihydroxy-1,4-benzoquinone dianion, and squarate as bridging ligands in the synthesis and magnetic properties of binuclear iron(III) complexes with oxalate. A Dinuclear 2,5-Dihydroxy-1,4-benzoquinone acts as a bridging ligand Ruthenium(II) Complex with the Dianion.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
615-94-1 | |
140.09 | |
QFSYADJLNBHAKO-UHFFFAOYSA-N | |
69213 | |
OC1=CC(=O)C(O)=CC1=O |
C6H4O4 | |
MFCD00001598 | |
2,5-dihydroxy-1,4-benzoquinone, 2,5-dihydroxy-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dihydroxy, p-benzoquinone, 2,5-dihydroxy, 2,5-dihydroxybenzoquinone, 4lbp, 2,5-dihydroxy-p-quinone, 2,5-dihydroxy benzoquinone, 2,5-dhbqop | |
2,5-dihydroxycyclohexa-2,5-diene-1,4-dione |
Specifications
615-94-1 | |
C6H4O4 | |
1939229 | |
Soluble in water. | |
OC1=CC(=O)C(O)=CC1=O | |
140.09 | |
140.1 | |
2,5-Dihydroxy-1,4-benzoquinone |
∼220°C (sublimation) | |
MFCD00001598 | |
2,5-dihydroxy-1,4-benzoquinone, 2,5-dihydroxy-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dihydroxy, p-benzoquinone, 2,5-dihydroxy, 2,5-dihydroxybenzoquinone, 4lbp, 2,5-dihydroxy-p-quinone, 2,5-dihydroxy benzoquinone, 2,5-dhbqop | |
QFSYADJLNBHAKO-UHFFFAOYSA-N | |
2,5-dihydroxycyclohexa-2,5-diene-1,4-dione | |
69213 | |
98% |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 210-454-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only