missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxy-1,4-benzoquinone, 98%, Thermo Scientific Chemicals

Product Code. 10666392
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-94-1
Molecular Formula C6H4O4
Molecular Weight (g/mol) 140.09
MDL Number MFCD00001598
InChI Key QFSYADJLNBHAKO-UHFFFAOYSA-N
Synonym 2,5-dihydroxy-1,4-benzoquinone, 2,5-dihydroxy-p-benzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dihydroxy, p-benzoquinone, 2,5-dihydroxy, 2,5-dihydroxybenzoquinone, 4lbp, 2,5-dihydroxy-p-quinone, 2,5-dihydroxy benzoquinone, 2,5-dhbqop
PubChem CID 69213
IUPAC Name 2,5-dihydroxycyclohexa-2,5-diene-1,4-dione
SMILES OC1=CC(=O)C(O)=CC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 220°C
Color Ochre to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (HO)2C6H2(=O)2
Quantity 25 g
Beilstein 08, 377
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetic acid;,slightly soluble in acetone,dmso;,insoluble in diethyl ether
Formula Weight 140.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2, 5-Dihydroxy-1, 4-benzoquinone, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Dihydroxy-1,4-benzoquinone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.