missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dihydroxyterephthalic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in hot dimethylformamide.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.
Solubility
Soluble in hot dimethylformamide.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Specifications
Melting Point | >300°C |
Color | Gray to Green |
Quantity | 25 g |
Beilstein | 2696052 |
Solubility Information | Soluble in hot dimethylformamide. |
Formula Weight | 198.13 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 2,5-Dihydroxyterephthalic acid |
Safety and Handling
Product Identifier
- 2,5-Dihydroxyterephthalic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : 2,5-Dihydroxyterephthalic acid
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'