missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt, 97%, Thermo Scientific Chemicals Product Code.: 11357556

2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt, 97%, Thermo Scientific Chemicals

Product Code. 11357556
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11357556

Brand: Thermo Scientific Alfa Aesar L11105.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1,3,4-thiadiazole-2,5-dithiol (Bismuthiol I) was used in determination of tellurium in indium phosphide doped with cadmium telluride by electro thermal atomic absorption spectrometry. It forms polymers with metals and can be useful in treatment of metal-polluted water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3,4-thiadiazole-2,5-dithiol (Bismuthiol I) was used in determination of tellurium in indium phosphide doped with cadmium telluride by electro thermal atomic absorption spectrometry. It forms polymers with metals and can be useful in treatment of metal-polluted water.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4628-94-8
Molecular Formula C2K2N2S3
Molecular Weight (g/mol) 226.413
MDL Number MFCD00003104
InChI Key GPWLFGDMYSVEGN-UHFFFAOYSA-L
Synonym 2,5-dimercapto-1,3,4-thiadiazole, dipotassium salt, dipotassium 1,3,4-thiadiazole-2,5-dithiolate, dipotassium 1,3,4-thiadiazole-2,5-bis thiolate, 1,3,4-thiadiazolidine-2,5-dithione, dipotassium salt, 1,3,4-thiadiazolidine-2,5-dithione, potassium salt 1:2, dipotassium 5-sulfanidyl-1,3,4-thiadiazol-2-yl sulfanide, 2,5-dimercapto-1,3,4-thiadiazole dipotassium salt, 1,3,4-thiadiazole-2,5-dithiol dipotassium salt, 2,5-di potassiothio-1,3,4-thiadiazole
PubChem CID 78364
IUPAC Name dipotassium;1,3,4-thiadiazole-2,5-dithiolate
SMILES C1(=NN=C(S1)[S-])[S-].[K+].[K+]

Specifications

Melting Point ∽274°C (decomposition)
Quantity 5 g
Beilstein 4917786
Solubility Information Insoluble in water.
Formula Weight 226.43
Percent Purity 97%
Chemical Name or Material 2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dimercapto-1,3,4-thiadiazole dipotassium salt, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.