missing translation for 'onlineSavingsMsg'
Learn More
2,5-Dimethoxy-2,5-dihydrofuran, 97%, mixture of cis and trans, Thermo Scientific Chemicals Product Code.: 13408318

2,5-Dimethoxy-2,5-dihydrofuran, 97%, mixture of cis and trans, Thermo Scientific Chemicals

Product Code. 13408318
100 g, Glass bottle
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 13408318

Brand: Thermo Scientific Acros 115531000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 332-77-4
CAS Min % 98.5
CAS Max % 100.0
Density 1.0700g/mL
Boiling Point 160.0°C to 162.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C6H10O3
Refractive Index 1.4330 to 1.4360
MDL Number MFCD00003220
Quantity 100 g
Specific Gravity 1.07
Synonym 2,5-dihydro-2,5-dimethoxyfuran, furan, 2,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran, cis+trans, 2,5-dihydro-2,5-dimethoxyfuran, mixture of cis and trans, pubchem6933, ksc490q3d, furan,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran,c&t, 2,5-dimethoxy-2,5-dihydrofuran,c&t, 2,5-dihydro-2,5-dimethoxyfuran, cis + trans
Solubility Information Solubility in water: 368g/L (20°C)
InChI Key WXFWXFIWDGJRSC-UHFFFAOYNA-N
SMILES COC1OC(OC)C=C1
IUPAC Name 2,5-dimethoxy-2,5-dihydrofuran
Molecular Weight (g/mol) 130.14
PubChem CID 78974
Viscosity 2.1 mPa.s (20°C)
Formula Weight 130.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2,5-Dimethoxy-2,5-dihydrofuran, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462739 Certificate TypeCertificate of Analysis Date17/05/2024 Catalog Number115531000
Lot NumberA0455504 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number115531000
Lot NumberA0369370 Certificate TypeCertificate of Analysis Date01/02/2022 Catalog Number115531000
compliance-icons compliance-icons
Product Identifier
  • 2,5-Dimethoxy-2,5-dihydrofuran,mixture of cis and trans
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,5-Dimethoxy-2,5-dihydrofuran, 97%, mixture of cis and trans, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.