missing translation for 'onlineSavingsMsg'
Learn More

2',5'-Dimethoxyacetophenone, 99%, Thermo Scientific Chemicals

18.60 € - 142.00 €

Chemical Identifiers

CAS 1201-38-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00008728
InChI Key FAXUIYJKGGUCBO-UHFFFAOYSA-N
Synonym 2',5'-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethanone, 2,5-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethan-1-one, ethanone, 1-2,5-dimethoxyphenyl, 1-2,5-dimethoxyphenyl-ethanone, 2-acetyl-1,4-dimethoxybenzene, acetophenone, 2',5'-dimethoxy, 1-acetyl-2,5-dimethoxybenzene, pubchem13430
PubChem CID 70991
IUPAC Name 1-(2,5-dimethoxyphenyl)ethanone
SMILES COC1=CC=C(OC)C(=C1)C(C)=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10608444
View Documents Promotion Details
Thermo Scientific Acros
115340050
5 g Glass bottle
18.60 €
5g
Please to purchase this item. Need a web account? Register with us today!
10080820
View Documents Promotion Details
Thermo Scientific Acros
115340250
25 g Glass bottle
51.00 €
25g
Please to purchase this item. Need a web account? Register with us today!
10795751
View Documents Promotion Details
Thermo Scientific Acros
115341000
100 g Glass bottle
142.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1201-38-3
180.20
FAXUIYJKGGUCBO-UHFFFAOYSA-N
70991
COC1=CC=C(OC)C(=C1)C(C)=O
C10H12O3
MFCD00008728
2',5'-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethanone, 2,5-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethan-1-one, ethanone, 1-2,5-dimethoxyphenyl, 1-2,5-dimethoxyphenyl-ethanone, 2-acetyl-1,4-dimethoxybenzene, acetophenone, 2',5'-dimethoxy, 1-acetyl-2,5-dimethoxybenzene, pubchem13430
1-(2,5-dimethoxyphenyl)ethanone

Specifications

1201-38-3
100.0
Green-Yellow to Yellow
155°C to 158°C (11.0 mmHg)
Authentic
C10H12O3
1.5421 to 1.5441
08, 271
2',5'-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethanone, 2,5-dimethoxyacetophenone, 1-2,5-dimethoxyphenyl ethan-1-one, ethanone, 1-2,5-dimethoxyphenyl, 1-2,5-dimethoxyphenyl-ethanone, 2-acetyl-1,4-dimethoxybenzene, acetophenone, 2',5'-dimethoxy, 1-acetyl-2,5-dimethoxybenzene, pubchem13430
COC1=CC=C(OC)C(=C1)C(C)=O
180.20
180.2
Liquid
98.5
18°C to 20°C
1.1300g/mL
>112°C
98.5% min. (GC)
(CH3O)2C6H3COCH3
MFCD00008728
1.13
FAXUIYJKGGUCBO-UHFFFAOYSA-N
1-(2,5-dimethoxyphenyl)ethanone
70991
99%
2', 5'-Dimethoxyacetophenone, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 214-858-0

missing translation for 'tsca' : TSCA

Product Identifier
  • 2',5'-Dimethoxyacetophenone

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only