missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethoxyaniline, 99%, Thermo Scientific Chemicals

Product Code. 11381807
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-56-7
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00008368
InChI Key NAZDVUBIEPVUKE-UHFFFAOYSA-N
Synonym benzenamine, 2,5-dimethoxy, aminohydroquinone dimethyl ether, 1-amino-2,5-dimethoxybenzene, 2,5-dimethoxybenzenamine, aniline, 2,5-dimethoxy, unii-v3z5u3fl10, 2,5 dimethoxyaniline, 2,5-dimethoxyphenylamine, dimethoxyaniline 2,5-, 2,5-dimethoxy aniline
PubChem CID 7613
IUPAC Name 2,5-dimethoxyaniline
SMILES COC1=CC=C(OC)C(N)=C1

Specifications

Melting Point 78°C to 82°C
Boiling Point 140°C (7 mmHg)
Flash Point 130°C (266°F)
Odor Characteristic
Quantity 50 g
UN Number UN2811
Beilstein 776823
Formula Weight 153.18
Percent Purity 99%
Chemical Name or Material 2,5-Dimethoxyaniline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'