missing translation for 'onlineSavingsMsg'
Learn More

2,5-dimethoxyphenyl isothiocyanate, Thermo Scientific™

Product Code. 10037592
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 40532-06-7
Molecular Formula C9H9NO2S
Molecular Weight (g/mol) 195.24
MDL Number MFCD00041059
InChI Key RADQLZMDTYWZKA-UHFFFAOYSA-N
Synonym 2,5-dimethoxyphenyl isothiocyanate, 2,5-dimethoxyphenylisothiocyanate, 2-isothiocyanato-1,4-dimethoxy-benzene, benzene, 2-isothiocyanato-1,4-dimethoxy, 2,5-dimethoxybenzenisothiocyanate, acmc-1amrr, #
PubChem CID 142466
IUPAC Name 2-isothiocyanato-1,4-dimethoxybenzene
SMILES COC1=CC=C(OC)C(=C1)N=C=S

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 195.24
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethoxyphenyl isothiocyanate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.