missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[((5-[(dimethylamino)methyl]-2-furyl)methyl)thio]ethan-1-amine, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Assay Percent Range | 95% |
Quantity | 1 g |
Formula Weight | 214.33 |
Percent Purity | 95% |
Chemical Name or Material | 2-[({5-[(Dimethylamino)methyl]-2-furyl}methyl)thio]ethan-1-amine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'