Learn More
2,5-Dimethylaniline, 98+%, Thermo Scientific Chemicals
Description
2,5-Dimethylaniline is widely used as raw materials to produce imaging chemicals like organic dyes and pigments. It is also used in the production of antioxidants, pharmaceutical, agricultural, rubber chemicals and other target organic molecules. 2,5-Dimethylaniline was also used in the synthesis of nanocomposite of multi-walled carbon nanotubes embedded in poly(2,5-dimethylaniline). 2,5-Dimethylaniline was used to study the effect of metabolites formed from 2,5-xylidine by fungi on laccase activity. The Fourier transform infrared (FTIR) and Raman spectra of 2,5-dimethylaniline was studied5. It induced transcription of lccIV.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2,5-Dimethylaniline is widely used as raw materials to produce imaging chemicals like organic dyes and pigments. It is also used in the production of antioxidants, pharmaceutical, agricultural, rubber chemicals and other target organic molecules. 2,5-Dimethylaniline was also used in the synthesis of nanocomposite of multi-walled carbon nanotubes embedded in poly(2,5-dimethylaniline). 2,5-Dimethylaniline was used to study the effect of metabolites formed from 2,5-xylidine by fungi on laccase activity. The Fourier transform infrared (FTIR) and Raman spectra of 2,5-dimethylaniline was studied5. It induced transcription of lccIV.
Solubility
Soluble in water <0.1 g/100 mL at 18°C.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
Specifications
Specifications
Melting Point | 15°C |
Boiling Point | 218°C |
Quantity | 25 g |
Odor | Characteristic |
UN Number | UN1711 |
Beilstein | 2205178 |
Merck Index | 14,10084 |
Formula Weight | 121.18 |
Percent Purity | ≥98% |
Chemical Name or Material | 2,5-Dimethylaniline |
RUO – Research Use Only