missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylphenylhydrazine hydrochloride, 98%, Thermo Scientific™

Product Code. 11363186
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56737-78-1
Molecular Formula C8H13ClN2
Molecular Weight (g/mol) 172.656
MDL Number MFCD00013382
InChI Key OMOFMSZLYCEIAF-UHFFFAOYSA-N
Synonym 2,5-dimethylphenylhydrazine hydrochloride, 2,5-dimethylphenyl hydrazine hydrochloride, 2,5-dimethylphenylhydrazinehydrochloride, 2,3-dimethylphenylhydrazinehydrochloride, pubchem6400, acmc-1bf0t, ksc491e8b, 2-hydrazino-m-xylene hydrochloride, 2,5-dimethyl-phenylhydrazine hydrochloride, 1-2,5-dimethylphenyl hydrazine hydrochloride
PubChem CID 2734058
IUPAC Name (2,5-dimethylphenyl)hydrazine;hydrochloride
SMILES CC1=CC(=C(C=C1)C)NN.Cl

Specifications

Melting Point ∽205°C (decomposition)
Quantity 25 g
Beilstein 6118180
Formula Weight 172.66
Percent Purity 98%
Chemical Name or Material 2,5-Dimethylphenylhydrazine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'