Learn More
2,5-Dimethylpyrrole, 97%, Thermo Scientific Chemicals
36.78 € - 115.58 €
Chemical Identifiers
CAS | 625-84-3 |
---|---|
Molecular Formula | C6H9N |
Molecular Weight (g/mol) | 95.14 |
MDL Number | MFCD00005223 |
InChI Key | PAPNRQCYSFBWDI-UHFFFAOYSA-N |
Synonym | 2,5-dimethylpyrrole, 2,5-dimethylazole, 1h-pyrrole, 2,5-dimethyl, pyrrole, 2,5-dimethyl, unii-mz3oyf5521, 2,5-dimethyl-pyrrole, pyrrole,2,5-dimethyl, 2,5-dimethyl pyrrole, pyrrole, 2,5-dimethyl-8ci, 2,5-bimethylpyrrole |
PubChem CID | 12265 |
IUPAC Name | 2,5-dimethyl-1H-pyrrole |
SMILES | CC1=CC=C(N1)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10596013
|
Thermo Scientific
116750050 |
5 g |
36.78 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10351614
|
Thermo Scientific Acros
116750250 |
25 g |
115.58 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
625-84-3 | |
95.14 | |
PAPNRQCYSFBWDI-UHFFFAOYSA-N | |
12265 | |
CC1=CC=C(N1)C |
C6H9N | |
MFCD00005223 | |
2,5-dimethylpyrrole, 2,5-dimethylazole, 1h-pyrrole, 2,5-dimethyl, pyrrole, 2,5-dimethyl, unii-mz3oyf5521, 2,5-dimethyl-pyrrole, pyrrole,2,5-dimethyl, 2,5-dimethyl pyrrole, pyrrole, 2,5-dimethyl-8ci, 2,5-bimethylpyrrole | |
2,5-dimethyl-1H-pyrrole |
Specifications
625-84-3 | |
100.0 | |
0.9350g/mL | |
54°C | |
96% min. (GC) | |
C6H9N | |
MFCD00005223 | |
0.935 | |
Solubility in water: insoluble | |
CC1=CC=C(N1)C | |
95.14 | |
95.14 | |
Crystalline Powder and/or Chunks |
96.0 | |
Orange-Yellow to Yellow | |
165.0°C (740.0 mmHg) | |
Authentic | |
Glass bottle | |
1.5040 to 1.5070 | |
20, 172 | |
2,5-dimethylpyrrole, 2,5-dimethylazole, 1h-pyrrole, 2,5-dimethyl, pyrrole, 2,5-dimethyl, unii-mz3oyf5521, 2,5-dimethyl-pyrrole, pyrrole,2,5-dimethyl, 2,5-dimethyl pyrrole, pyrrole, 2,5-dimethyl-8ci, 2,5-bimethylpyrrole | |
PAPNRQCYSFBWDI-UHFFFAOYSA-N | |
2,5-dimethyl-1H-pyrrole | |
12265 | |
97% | |
2, 5-Dimethylpyrrole |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-913-8
Recommended Storage : Product may darken during storage
- 2,5-Dimethylpyrrole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.