missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diphenyl-p-benzoquinone, 96%, Thermo Scientific Chemicals

Product Code. 11468306
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,5-Diphenyl-p-benzoquinone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Diphenyl-p-benzoquinone is used as pharmaceutical intermediate.

Solubility
Soluble in hot Toluene (almost transparency).

Notes
Incompatible with Oxidizing agents, Reducing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 844-51-9
Molecular Formula C18H12O2
Molecular Weight (g/mol) 260.29
MDL Number MFCD00001600
InChI Key QYXHDJJYVDLECA-UHFFFAOYSA-N
Synonym 2,5-diphenyl-1,4-benzoquinone, 2,5-diphenyl-p-benzoquinone, 2,5-diphenylquinone, 2,5-diphenyl-4-benzoquinone, 2,5-diphenylbenzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-diphenyl, 1,1':4',1-terphenyl-2',5'-dione, p-benzoquinone, 2,5-diphenyl, acmc-209pup, 2,5-diphenyl-p-quinone
PubChem CID 70055
IUPAC Name 2,5-diphenylcyclohexa-2,5-diene-1,4-dione
SMILES O=C1C=C(C(=O)C=C1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 215°C to 219°C
Quantity 25 g
Beilstein 2051812
Solubility Information Soluble in hot Toluene (almost transparency).
Formula Weight 260.3
Percent Purity 96%
Chemical Name or Material 2,5-Diphenyl-p-benzoquinone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'