missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Diphenyloxazole, Scintillation Grade 99%, Thermo Scientific Chemicals
32.96 € - 1550.72 €
Chemical Identifiers
CAS | 92-71-7 |
---|---|
Molecular Formula | C15H11NO |
Molecular Weight (g/mol) | 221.26 |
MDL Number | MFCD00005306 |
InChI Key | CNRNYORZJGVOSY-UHFFFAOYSA-N |
Synonym | PPO |
PubChem CID | 7105 |
IUPAC Name | 2,5-diphenyl-1,3-oxazole |
SMILES | O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10091090
|
Thermo Scientific Acros
117380250 |
25 g | Plastic Bottle |
32.96 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10102770
|
Thermo Scientific Acros
117381000 |
100 g | Plastic Bottle |
87.33 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10112770
|
Thermo Scientific Acros
117385000 |
500 g | Plastic Bottle |
322.61 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10793324
|
Thermo Scientific Acros
117380025 |
2.5 kg | Plastic Drum |
1550.72 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for fluorescence/luminescence spectroscopyChemical Identifiers
92-71-7 | |
221.26 | |
CNRNYORZJGVOSY-UHFFFAOYSA-N | |
7105 | |
O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1 |
C15H11NO | |
MFCD00005306 | |
PPO | |
2,5-diphenyl-1,3-oxazole |
Specifications
70°C to 72°C | |
Crystalline Powder | |
100.0 | |
1.06g/mL | |
99% | |
C15H11NO | |
27, 78 | |
CNRNYORZJGVOSY-UHFFFAOYSA-N | |
2,5-diphenyl-1,3-oxazole | |
7105 | |
≥98.5% |
92-71-7 | |
98.5 | |
< 25 cpm | |
White | |
Authentic | |
MFCD00005306 | |
PPO | |
O1C(=CN=C1C1=CC=CC=C1)C1=CC=CC=C1 | |
221.26 | |
221.26 | |
2, 5-Diphenyloxazole, 99% |
Safety and Handling
missing translation for 'einecsNumber' : 202-181-3
missing translation for 'rtecsNumber' : RP6825000
missing translation for 'tsca' : TSCA
Product Identifier
- 2,5-Diphenyloxazole
RUO – Research Use Only