missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 107367-98-6
Molecular Formula C12H11NO3
Molecular Weight (g/mol) 217.224
MDL Number MFCD00100005
InChI Key XEWJNPORMBGGKZ-UHFFFAOYSA-N
Synonym 2-5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 2-5-methyl-2-phenyloxazol-4-yl acetic acid, 5-methyl-2-phenyloxazol-4-yl acetic acid, maybridge1_002118, acmc-2098vv, 2-phenyl-5-methyl-4-oxazolyiacetic acid, 2-phenyl-5-methyl-4-oxazolylacetic acid, 2-phenyl-5-methyloxazol-4-ylacetic acid, 5-methyl-2-phenyl-1,3-oxazolyl acetic acid
PubChem CID 2775139
IUPAC Name 2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid
SMILES CC1=C(N=C(O1)C2=CC=CC=C2)CC(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10354272
View Documents Promotion Details
Thermo Scientific Maybridge
BTB09929DA
1 g N/A
N/A
10641473
View Documents Promotion Details
Thermo Scientific Maybridge
BTB09929EA
10 g N/A
N/A
10546965
View Documents Promotion Details
Thermo Scientific Maybridge
BTB09929EB
25 g N/A
N/A
Description

Description

Specifications

Chemical Identifiers

107367-98-6
217.224
XEWJNPORMBGGKZ-UHFFFAOYSA-N
2775139
CC1=C(N=C(O1)C2=CC=CC=C2)CC(=O)O
C12H11NO3
MFCD00100005
2-5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 2-5-methyl-2-phenyloxazol-4-yl acetic acid, 5-methyl-2-phenyloxazol-4-yl acetic acid, maybridge1_002118, acmc-2098vv, 2-phenyl-5-methyl-4-oxazolyiacetic acid, 2-phenyl-5-methyl-4-oxazolylacetic acid, 2-phenyl-5-methyloxazol-4-ylacetic acid, 5-methyl-2-phenyl-1,3-oxazolyl acetic acid
2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid

Specifications

107367-98-6
100.0
Amber Glass Bottle
MFCD00100005
XEWJNPORMBGGKZ-UHFFFAOYSA-N
2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid
2775139
97%
96.5
97%
C12H11NO3
2-5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 2-5-methyl-2-phenyloxazol-4-yl acetic acid, 5-methyl-2-phenyloxazol-4-yl acetic acid, maybridge1_002118, acmc-2098vv, 2-phenyl-5-methyl-4-oxazolyiacetic acid, 2-phenyl-5-methyl-4-oxazolylacetic acid, 2-phenyl-5-methyloxazol-4-ylacetic acid, 5-methyl-2-phenyl-1,3-oxazolyl acetic acid
CC1=C(N=C(O1)C2=CC=CC=C2)CC(=O)O
217.224
217.22
2-(5-Methyl-2-phenyl-1, 3-oxazol-4-yl)acetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications