missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Norbornadiene, 97%, stab with 250 ppm BHT, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
Description
2,5-Norbornadiene is used as an intermediate in prostaglandin synthesis and shows anti-ethylene effect on plants. It acts as a starting material for the synthesis of diamantane and as an acetylene transfer agent for instance, in reaction with 3,6-di-2-pyridyl-1,2,4,5-tetrazine. It is also a useful dienophile in Diels-Alder reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,5-Norbornadiene is used as an intermediate in prostaglandin synthesis and shows anti-ethylene effect on plants. It acts as a starting material for the synthesis of diamantane and as an acetylene transfer agent for instance, in reaction with 3,6-di-2-pyridyl-1,2,4,5-tetrazine. It is also a useful dienophile in Diels-Alder reactions.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
2,5-Norbornadiene is used as an intermediate in prostaglandin synthesis and shows anti-ethylene effect on plants. It acts as a starting material for the synthesis of diamantane and as an acetylene transfer agent for instance, in reaction with 3,6-di-2-pyridyl-1,2,4,5-tetrazine. It is also a useful dienophile in Diels-Alder reactions.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.
Specifications
Melting Point | -20°C |
Density | 0.906 |
Boiling Point | 88°C to 90°C |
Flash Point | −11°C (12°F) |
Refractive Index | 1.47 |
Quantity | 100 mL |
UN Number | UN2251 |
Beilstein | 506224 |
Solubility Information | Immiscible with water. |
Formula Weight | 92.14 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'