missing translation for 'onlineSavingsMsg'
Learn More

2,5-Pyridinedicarboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 10284750
Click to view available options
5g; Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 100-26-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 256°C
Color Yellow to Green
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H5NO4
Linear Formula (C5H3N)(COOH)2
MDL Number MFCD00006297
Quantity 5 g
Beilstein 22, 153
Merck Index 15, 5204
Synonym 2,5-pyridinedicarboxylic acid, isocinchomeronic acid, 2,5-pyridinedicarboxylicacid, unii-4k1kgr926x, pyridinedicarboxylic acid-2,5, chembl88972, pyridine 2,5-dicarboxylate, pubchem12991, 2,5-dicarboxy-pyridine, 6-carboxy-nicotinic acid
Solubility Information Solubility in water: insoluble in cold water. Other solubilities: soluble in hot water,insoluble in alcohol,ether and benzene,slightly soluble in boiling alcohol
InChI Key LVPMIMZXDYBCDF-UHFFFAOYSA-N
SMILES C1=CC(=NC=C1C(=O)O)C(=O)O
IUPAC Name pyridine-2,5-dicarboxylic acid
Molecular Weight (g/mol) 167.12
PubChem CID 7493
ChEBI CHEBI:46865
Formula Weight 167.12
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2, 5-Pyridinedicarboxylic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2,5-Pyridinedicarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.