missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(hydroxymethyl)-p-cresol, tech. 90%, Thermo Scientific Chemicals

41.91 € - 484.41 €

Chemical Identifiers

CAS 91-04-3
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.192
MDL Number MFCD00004619
InChI Key KUMMBDBTERQYCG-UHFFFAOYSA-N
Synonym 2,6-bis hydroxymethyl-4-methylphenol, 2,6-bis hydroxymethyl-p-cresol, 2,6-dimethylol-p-cresol, 2-hydroxy-5-methyl-1,3-phenylene dimethanol, 2,6-dimethylol-4-methylphenol, 2,6-di hydroxymethyl-p-cresol, 1,3-benzenedimethanol, 2-hydroxy-5-methyl, 2-hydroxy-5-methyl-1,3-benzenedimethanol, 3,5-bis hydroxymethyl-p-cresol, alpha1,alpha,2-trihydroxymesitylene
PubChem CID 7039
IUPAC Name 2,6-bis(hydroxymethyl)-4-methylphenol
SMILES CC1=CC(=C(C(=C1)CO)O)CO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11493607
View Documents
Thermo Scientific Alfa Aesar
A17369.14
25 g
41.91 €
25g
Please to purchase this item. Need a web account? Register with us today!
11403617
View Documents
Thermo Scientific Alfa Aesar
A17369.22
100 g
137.69 €
100g
Please to purchase this item. Need a web account? Register with us today!
11413617
View Documents
Thermo Scientific Alfa Aesar
A17369.36
500 g
484.41 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

91-04-3
168.192
KUMMBDBTERQYCG-UHFFFAOYSA-N
7039
CC1=CC(=C(C(=C1)CO)O)CO
C9H12O3
MFCD00004619
2,6-bis hydroxymethyl-4-methylphenol, 2,6-bis hydroxymethyl-p-cresol, 2,6-dimethylol-p-cresol, 2-hydroxy-5-methyl-1,3-phenylene dimethanol, 2,6-dimethylol-4-methylphenol, 2,6-di hydroxymethyl-p-cresol, 1,3-benzenedimethanol, 2-hydroxy-5-methyl, 2-hydroxy-5-methyl-1,3-benzenedimethanol, 3,5-bis hydroxymethyl-p-cresol, alpha1,alpha,2-trihydroxymesitylene
2,6-bis(hydroxymethyl)-4-methylphenol

Specifications

91-04-3
C9H12O3
1240237
KUMMBDBTERQYCG-UHFFFAOYSA-N
2,6-bis(hydroxymethyl)-4-methylphenol
7039
90%
2,6-Bis(hydroxymethyl)-p-cresol
119°C to 126°C
MFCD00004619
2,6-bis hydroxymethyl-4-methylphenol, 2,6-bis hydroxymethyl-p-cresol, 2,6-dimethylol-p-cresol, 2-hydroxy-5-methyl-1,3-phenylene dimethanol, 2,6-dimethylol-4-methylphenol, 2,6-di hydroxymethyl-p-cresol, 1,3-benzenedimethanol, 2-hydroxy-5-methyl, 2-hydroxy-5-methyl-1,3-benzenedimethanol, 3,5-bis hydroxymethyl-p-cresol, alpha1,alpha,2-trihydroxymesitylene
CC1=CC(=C(C(=C1)CO)O)CO
168.192
168.19
Technical
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H332-H315-H319-H335

missing translation for 'einecsNumber' : 202-036-4

missing translation for 'rtecsNumber' : CZ6440000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only