missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromonaphthalene, 99%, Thermo Scientific Chemicals

Product Code. 11352897
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,6-Dibromonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It reacts with hexylmagnesium bromide to form 2,6-Dihexyl-naphthalene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromonaphthalene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. It reacts with hexylmagnesium bromide to form 2,6-Dihexyl-naphthalene.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13720-06-4
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.97
MDL Number MFCD01026462
InChI Key PJZDEYKZSZWFPX-UHFFFAOYSA-N
Synonym naphthalene, 2,6-dibromo, 2,6-dibromo-naphthalene, 2,6-dibromo naphthalene, 2.6-dibromonaphthalene, 2,6-?dibromonaphthalene, acmc-1c7c1, ksc490o3h, 2,6-dibromonaphthalene
PubChem CID 640591
IUPAC Name 2,6-dibromonaphthalene
SMILES BrC1=CC2=CC=C(Br)C=C2C=C1

Specifications

Melting Point 162°C to 167°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 285.97
Percent Purity 99%
Chemical Name or Material 2,6-Dibromonaphthalene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'