missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromophenol, 99%, Thermo Scientific Chemicals

Product Code. 11473224
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,6-Dibromophenol is used in the production of 3,4,5-trimethoxybenzaldehyde and 2-(2,6-dibromophenoxy)ethylbromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromophenol is used in the production of 3,4,5-trimethoxybenzaldehyde and 2-(2,6-dibromophenoxy)ethylbromide.

Solubility
Soluble in ethanol and ether. Slightly soluble in water.

Notes
Incompatible with acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-33-3
Molecular Formula C6H4Br2O
Molecular Weight (g/mol) 251.91
MDL Number MFCD00002152
InChI Key SSIZLKDLDKIHEV-UHFFFAOYSA-N
Synonym phenol, 2,6-dibromo, 2,6-dibromo-phenol, unii-27mip05eav, 2,6-dibromo-pheno, 27mip05eav, 2,6-bis bromanyl phenol, acmc-1au9i, 2,6-dibromophenol, ksc493e5t, 2,6-dibromophenol, analytical standard
PubChem CID 11847
ChEBI CHEBI:19391
IUPAC Name 2,6-dibromophenol
SMILES OC1=C(Br)C=CC=C1Br

Specifications

Melting Point 53°C to 57°C
Boiling Point 255°C to 256°C
Odor Phenol-like
Quantity 1 g
Beilstein 2043614
Solubility Information Soluble in ethanol and ether. Slightly soluble in water.
Formula Weight 251.92
Percent Purity 99%
Chemical Name or Material 2,6-Dibromophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'