missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromopyrazine, 95%, Thermo Scientific Chemicals

Product Code. 15488018
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,6-Dibromopyrazine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Dibromopyrazine is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Storage temperature 2 - 4°Cand Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23229-25-6
Molecular Formula C4H2Br2N2
Molecular Weight (g/mol) 237.88
MDL Number MFCD09834804
InChI Key JXKQTRCEKQCAGH-UHFFFAOYSA-N
Synonym pyrazine, 2,6-dibromo, 2,6-dibromo-pyrazine, acmc-1cj6s, ksc201c7f
PubChem CID 20361700
IUPAC Name 2,6-dibromopyrazine
SMILES BrC1=CN=CC(Br)=N1

Specifications

Color White to Brown
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 237.88
Percent Purity 95%
Physical Form Crystalline Solid
Chemical Name or Material 2,6-Dibromopyrazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.