missing translation for 'onlineSavingsMsg'
Learn More
2,6-Dichloro-3-nitrobenzonitrile, 98%, Thermo Scientific Chemicals Product Code.: 11372406

2,6-Dichloro-3-nitrobenzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11372406
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11372406

Brand: Thermo Scientific Alfa Aesar L06186.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5866-98-8
Molecular Formula C7H2Cl2N2O2
Molecular Weight (g/mol) 217.005
MDL Number MFCD00051513
InChI Key NSKVWZIEYFSHIM-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-nitrobenzenecarbonitrile, benzonitrile, 2,6-dichloro-3-nitro, acmc-1ala2, dichloronitrobenzenecarbonitrile, 2,6-dichloro-3-nitorbenzonitirle, 2,6-bis chloranyl-3-nitro-benzenecarbonitrileShow MoreShow Less
PubChem CID 4461932
IUPAC Name 2,6-dichloro-3-nitrobenzonitrile
SMILES C1=CC(=C(C(=C1[N+](=O)[O-])Cl)C#N)Cl

Specifications

Melting Point 106°C to 109°C
Quantity 5 g
UN Number UN3439
Beilstein 2113045
Formula Weight 217.01
Percent Purity 98%
Chemical Name or Material 2,6-Dichloro-3-nitrobenzonitrile

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10155494 Certificate TypeCertificate of Analysis Date22/04/2024 Catalog NumberL06186.06
Lot Number10143207 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL06186.06
Lot Number10150322 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL06186.06
Lot NumberGBFA033623 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL06186.06
Lot Number10120247 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL06186.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,6-Dichloro-3-nitrobenzonitrile, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.