missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichloro-3-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 15494597
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25922-41-2
Molecular Formula C8H3Cl2F3O2
Molecular Weight (g/mol) 259.01
MDL Number MFCD09743917
InChI Key WRNSEKNHEIXBES-UHFFFAOYSA-N
Synonym 2,6-dichloro-3-trifluoromethyl benzoic acid, 2,6-dichloro-3-trifluoromethyl-benzoic acid, 2,6-dichloro-3-trifluoromethyl benzoic acid, jrd
PubChem CID 11680459
IUPAC Name 2,6-dichloro-3-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=C(Cl)C=CC(=C1Cl)C(F)(F)F

Specifications

Melting Point 98°C to 102°C
Quantity 5 g
Formula Weight 259.01
Percent Purity 97%
Chemical Name or Material 2,6-Dichloro-3-(trifluoromethyl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'