missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dichloro-4-(chloromethyl)pyridine, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 5 g |
Formula Weight | 196.46 |
Percent Purity | 95% |
Chemical Name or Material | 2, 6-Dichloro-4-(chloromethyl)pyridine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu