missing translation for 'onlineSavingsMsg'
Learn More

(2,6-Dichloro-4-pyridyl)methanol, 95%, Thermo Scientific™

Product Code. 10166962
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 101990-69-6
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.012
MDL Number MFCD00052638
InChI Key YDGJTFCKLFLWFM-UHFFFAOYSA-N
Synonym 2,6-dichloropyridine-4-methanol, 2,6-dichloropyridin-4-yl methanol, 2,6-dichloro-4-hydroxymethyl pyridine, 2,6-dichloro-4-pyridyl methanol, 4-pyridinemethanol, 2,6-dichloro, 4-pyridinemethanol,2,6-dichloro, 2,6-dichloro-4-pyridinyl methanol, 2,6-dichloro-pyridin-4-yl-methanol, 2,6-dichloro-4-pyridinemethanol, acmc-209zae
PubChem CID 2794840
IUPAC Name (2,6-dichloropyridin-4-yl)methanol
SMILES C1=C(C=C(N=C1Cl)Cl)CO

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 178.02
Percent Purity 95%
Chemical Name or Material (2, 6-Dichloro-4-pyridyl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'