missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2,6-Dichloro-4-pyridyl)methanol, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 1 g |
Formula Weight | 178.02 |
Percent Purity | 95% |
Chemical Name or Material | (2, 6-Dichloro-4-pyridyl)methanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'