missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzaldehyde, 99%, Thermo Scientific Chemicals

34.47 € - 491.45 €

Chemical Identifiers

CAS 83-38-5
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00003307
InChI Key DMIYKWPEFRFTPY-UHFFFAOYSA-N
Synonym benzaldehyde, 2,6-dichloro, ccris 6014, 2,6-dichloro benzaldehyde, 2,6-dichloro-benzaldehyde, 2,6-dichloro-benzaldehyd, 2,6-dichlorbenzaldehyd, pubchem8225, acmc-1bkey, 2,6dichloro-benzaldehyde, benzaldehyde,6-dichloro
PubChem CID 6737
IUPAC Name 2,6-dichlorobenzaldehyde
SMILES C1=CC(=C(C(=C1)Cl)C=O)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10510201
View Documents
Thermo Scientific Acros
113160500
50 g Glass bottle
34.47 €
50g
Please to purchase this item. Need a web account? Register with us today!
10408640
View Documents
Thermo Scientific Acros
113162500
250 g Glass bottle
129.65 €
250g
Please to purchase this item. Need a web account? Register with us today!
10279693
View Documents
Thermo Scientific Acros
113160010
1 kg Glass bottle
491.45 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

83-38-5
175.01
DMIYKWPEFRFTPY-UHFFFAOYSA-N
6737
C1=CC(=C(C(=C1)Cl)C=O)Cl
C7H4Cl2O
MFCD00003307
benzaldehyde, 2,6-dichloro, ccris 6014, 2,6-dichloro benzaldehyde, 2,6-dichloro-benzaldehyde, 2,6-dichloro-benzaldehyd, 2,6-dichlorbenzaldehyd, pubchem8225, acmc-1bkey, 2,6dichloro-benzaldehyde, benzaldehyde,6-dichloro
2,6-dichlorobenzaldehyde

Specifications

83-38-5
100.0
White to Beige
Authentic
C7H4Cl2O
MFCD00003307
benzaldehyde, 2,6-dichloro, ccris 6014, 2,6-dichloro benzaldehyde, 2,6-dichloro-benzaldehyde, 2,6-dichloro-benzaldehyd, 2,6-dichlorbenzaldehyd, pubchem8225, acmc-1bkey, 2,6dichloro-benzaldehyde, benzaldehyde,6-dichloro
DMIYKWPEFRFTPY-UHFFFAOYSA-N
2,6-dichlorobenzaldehyde
6737
99%
2, 6-Dichlorobenzaldehyde, 99%
98.5
71.0°C to 72.0°C
>100°C
99%
Cl2C6H3CHO
07, 237
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and ligroin
C1=CC(=C(C(=C1)Cl)C=O)Cl
175.01
175.01
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 201-472-2

missing translation for 'tsca' : TSCA

Product Identifier
  • 2,6-Dichlorobenzaldehyde

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.